VWR®, 使い捨てマイクロ秤量皿
0,12 ml, 12×4×2,7 mm
Catalog # 611-1360
Supplier: VWR Collection
Jump to:
Product Details & Documents
Aluminium.
These micro weighing boats are compatible with most commercially available analysers and microbalances, and are packed in easy to use plastic containers for storage and dispensing.
Documents
Specifications
- 容量:0,12 ml
- Seq:09
- W×D×H:12×4×2,7 mm