Orders LinkContactUsLinkComponent
VWR®, 使い捨てマイクロ秤量皿
0,04 ml, 6,5×3,5×2,5 mm
Catalog # 611-1357
Supplier:   VWR Collection
VWR®, 使い捨てマイクロ秤量皿
Catalog # 611-1357
Supplier:  VWR Collection
VWR®, 使い捨てマイクロ秤量皿
VWR®, 使い捨てマイクロ秤量皿
0,04 ml, 6,5×3,5×2,5 mm
Price
Inquire For Price
Log in for availability
1 * 250 items
Product Details & Documents
Aluminium.
These micro weighing boats are compatible with most commercially available analysers and microbalances, and are packed in easy to use plastic containers for storage and dispensing.
Specifications
  • 容量:
    0,04 ml
  • Seq:
    05
  • W×D×H:
    6,5×3,5×2,5 mm