VWR®, 使い捨てマイクロ秤量皿
0,35 ml, 13×3,5 mm
611-1356
Aluminium.
These micro weighing boats are compatible with most commercially available analysers and microbalances, and are packed in easy to use plastic containers for storage and dispensing.
- Int.Ø×H:13×3,5 mm
- 容量:0,35 ml
- Seq:03