VWR®, 使い捨てマイクロ秤量皿
0,035 ml, 6×2,5 mm
611-1355
Aluminium.
These micro weighing boats are compatible with most commercially available analysers and microbalances, and are packed in easy to use plastic containers for storage and dispensing.
- Int.Ø×H:6×2,5 mm
- 容量:0,035 ml
- Seq:01