Orders LinkContactUsLinkComponent
VWR®, 校正用分銅、OIMLクラスF2
10 kg, 150 ± mg
  611-8176
VWR®, 校正用分銅、OIMLクラスF2
  611-8176
VWR®, 校正用分銅、OIMLクラスF2
VWR®, 校正用分銅、OIMLクラスF2
10 kg, 150 ± mg
1 * 1 items
Stainless steel Class F2 weights for users who require moderate levels of tolerance values in their calibration and day-to-day weighing.
  • Wire weights up to 500 mg in cobalt alloy
  • Identification number guarantees traceability for weights from 1 g upwards
  • Laser engraving improves legibility of the markings
  • Available with or without COFRAC certificate
 : Conform to OILM R111 recommendations.
 : All weights from 1 mg to 20 kg are available with or without COFRAC certificate - please visit website or contact your local Avantor sales office. A range of sets of weights are also available - please search for product 'Calibration weight sets, Class F2'.
 : Plastic cases have a vacuum sealed protective film to ensure that the case and calibration weight are protected during transportation.
  • Weight:
    10 kg
  • Seq:
    29
  • Tolerance:
    150 ± mg