Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 校正用分銅、OIMLクラスF2

VWR®, 校正用分銅、OIMLクラスF2

5 kg, 75 ± mg
Catalog # 611-8175
Supplier:   VWR Collection
VWR®, 校正用分銅、OIMLクラスF2
Catalog # 611-8175
Supplier:  VWR Collection
VWR®, 校正用分銅、OIMLクラスF2
VWR®, 校正用分銅、OIMLクラスF2

5 kg, 75 ± mg

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Stainless steel Class F2 weights for users who require moderate levels of tolerance values in their calibration and day-to-day weighing.

  • Wire weights up to 500 mg in cobalt alloy
  • Identification number guarantees traceability for weights from 1 g upwards
  • Laser engraving improves legibility of the markings
  • Available with or without COFRAC certificate

Certifications: Conform to OILM R111 recommendations.

注文情報: All weights from 1 mg to 20 kg are available with or without COFRAC certificate - please visit website or contact your local Avantor sales office. A range of sets of weights are also available - please search for product 'Calibration weight sets, Class F2'.

包装: Plastic cases have a vacuum sealed protective film to ensure that the case and calibration weight are protected during transportation.

Specifications
  • Weight:
    5 kg
  • Seq:
    28
  • Tolerance:
    75 ± mg
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.