レフィル用セット, SPILL-X®
Formaldehyde refill, SPILL-X-FP®, 6 bottle set
Catalog # 121-0013
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
These refill sets can be used with five types of SPILL-X® treatment kits. They are available in different formats.
- Each set contains six bottles
納品情報: Multipurpose SPILL-X® set contains 2 acids, 2 caustics and 2 solvent agents, whereas the formaldehyde SPILL-X-FP® refill set comprises 6 bottles of formaldehyde agents.
Specifications
- 説明:Formaldehyde refill, SPILL-X-FP®, 6 bottle set