Orders LinkContactUsLinkComponent
レフィル用セット, SPILL-X®
レフィル用セット, SPILL-X®
Solvent refill, SPILL-X®, 6 bottle set
Catalog # 121-0012
Supplier:   VWR Chemicals
レフィル用セット, SPILL-X®
Catalog # 121-0012
Supplier:  VWR Chemicals
レフィル用セット, SPILL-X®
レフィル用セット, SPILL-X®
Solvent refill, SPILL-X®, 6 bottle set
Price
Inquire For Price
Log in for availability
1 * 1 SET
Product Details & Documents
These refill sets can be used with five types of SPILL-X® treatment kits. They are available in different formats.
  • Each set contains six bottles
納品情報: Multipurpose SPILL-X® set contains 2 acids, 2 caustics and 2 solvent agents, whereas the formaldehyde SPILL-X-FP® refill set comprises 6 bottles of formaldehyde agents.
Specifications
  • 説明:
    Solvent refill, SPILL-X®, 6 bottle set