VWR®, 特殊pH電極
Flat tip, double junction (HDPE/Pellon), 10...100 °C, Glass, 3,5 M KCl, 12×150 mm, BNC, Agar, food and beverage, serum, soil, Tris buffers, viscous samples
Supplier: VWR Collection
Jump to:
Product Details & Documents
pH electrodes with 1 m fixed cable or S7 connector.
- Checked at five points during the manufacturing process and lot stamped to ensure quality
- Storage soaker bottle to protect the electrode, keeping the item in top condition and instantly ready for use
pH range: 0 to 14
Temperature range: 0 to +100 °C (10 to 100 °C for flat tips)
Response time: 95% <1 second (95% <5 seconds for flat tips)
Documents
Specifications
- Connection:BNC
- 直径×長さ:12×150 mm
- For:Agar, food and beverage, serum, soil, Tris buffers, viscous samples
- Type:Flat tip, double junction (HDPE/Pellon)
- Material:Glass
- Seq:17
- Electrolyte:3,5 M KCl
- Temp.:10...100 °C