Orders LinkContactUsLinkComponent
酵母ナイトロジェンベース(YNB)、VWR®
酵母ナイトロジェンベース(YNB)、VWR®
Yeast nitrogen base without amino acids, 500 g
Catalog # J386-500G
Supplier:   VWR Chemicals
酵母ナイトロジェンベース(YNB)、VWR®
Catalog # J386-500G
Supplier:  VWR Chemicals
CAS Number:  
酵母ナイトロジェンベース(YNB)、VWR®
酵母ナイトロジェンベース(YNB)、VWR®
Yeast nitrogen base without amino acids, 500 g
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Yeast Nitrogen Base (YNB) is a base medium for preparation of minimal and synthetic defined yeast media.
  • Animal origin
  • Suitable for studying strains of yeast requiring certain vitamins
注文情報: Base materials are available with or without ammonium sulfate for the classification of yeast.
Documents
Specifications
Specifications
  • Pk:
    500 g
  • 説明:
    Yeast nitrogen base without amino acids
Specifications