酵母ナイトロジェンベース(YNB)、VWR®
Yeast nitrogen base without amino acids, 100 g
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Yeast Nitrogen Base (YNB) is a base medium for preparation of minimal and synthetic defined yeast media.
- Animal origin
- Suitable for studying strains of yeast requiring certain vitamins
Ordering information: Base materials are available with or without ammonium sulfate for the classification of yeast.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:100 g
- 説明:Yeast nitrogen base without amino acids