Orders LinkContactUsLinkComponent
LB培地(プレミックス)、VWR®製
LB broth, Miller formulation, tissue culture grade, 500 g
  J106-500G
LB培地(プレミックス)、VWR®製
  J106-500G
 :  
LB培地(プレミックス)、VWR®製
LB培地(プレミックス)、VWR®製
LB broth, Miller formulation, tissue culture grade, 500 g
 :  
1 * 500 g
Lysogeny broth (LB) is a nutritionally rich medium used for the propagation and maintenance of E. coli. It is also widely used for the preparation of plasmid DNA and recombinant proteins.
  • Save time using pre-mixed media
  • Homogenous and high quality powders
Formulation of 10 g tryptone, 10 g NaCl, and 5 g yeast extract (25 g powder total) per L of liquid broth.
The instructions included on the label state to prepare LB broth by dissolving 25 g powder per liter in deionized water.
 : Individual ReadyPack™ pouches prepare 1 L of media.
  • Pk:
    500 g
  • 説明:
    LB broth, Miller formulation, tissue culture grade