Sodium deoxycholate ≥99.0%, high purity
HDPE bottle, 50 g
Supplier: VWR Chemicals
Sodium deoxycholate ≥99.0%, high purity
Catalog # 0613-50G
Supplier: VWR Chemicals
CAS Number: 302-95-4
Jump to:
Product Details & Documents
Anionic - negatively-charged (carboxylates, sulphates or sulphonates in alkali metal form). Utilised for applications ranging from solubilising membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis. Used for complete disruption of membranes.
Synonyms: Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
- CAS Number:302-95-4
- Formula:C₂₄H₃₉NaO₄
- MDL Number:MFCD00064139
- Molecular Weight:414.56 g/mol
- Melting Pt:357…365 °C
- Storage Temperature:Ambient
- EINECS:206-132-7
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Arsenic:≤0.001%
- Heavy metals:≤0.005%
- Loss on drying:≤5.0%
- Purity (dry basis):≥99.0%
- Sodium cholate:≤2.0%
- Solubility (10%, water):Pass
Specifications
- パックタイプ:HDPE bottle
- Pk:50 g



