VWR®, 電極洗浄液
Electrode cleaning solution pepsin/hydrochloric acid for removal of proteins, Plastic bottle, 500 mL
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
For full information concerning health and safety data please see the website.
Specifications
Specifications
- パックタイプ:Plastic bottle
- Pk:500 mL
- 説明:Electrode cleaning solution pepsin/hydrochloric acid for removal of proteins