Orders LinkContactUsLinkComponent
VWR®, 電極洗浄液
VWR®, 電極洗浄液
Electrode cleaning solution pepsin/hydrochloric acid for removal of proteins, Plastic bottle, 500 mL
Catalog # 83603.260
Supplier:   VWR Chemicals
VWR®, 電極洗浄液
Catalog # 83603.260
Supplier:  VWR Chemicals
CAS Number:  
VWR®, 電極洗浄液
VWR®, 電極洗浄液
Electrode cleaning solution pepsin/hydrochloric acid for removal of proteins, Plastic bottle, 500 mL
CAS Number:  
Price
$97.64
Log in for availability
1 * 500 mL
Specifications
Specifications
  • パックタイプ:
    Plastic bottle
  • Pk:
    500 mL
  • 説明:
    Electrode cleaning solution pepsin/hydrochloric acid for removal of proteins
Specifications