Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, ピペットチップ、ユニバーサル

VWR®, ピペットチップ、ユニバーサル

2 - 200 µl, 50 mm, Yellow, Yellow, Bulk
Catalog # 612-5755
Supplier:   VWR Collection
VWR®, ピペットチップ、ユニバーサル
Catalog # 612-5755
Supplier:  VWR Collection
VWR®, ピペットチップ、ユニバーサル
VWR®, ピペットチップ、ユニバーサル

2 - 200 µl, 50 mm, Yellow, Yellow, Bulk

Price
Inquire For Price
Log in for availability
1 * 1.000 items
Product Details & Documents

PP, autoclavable.

  • Manufactured without lubricants
  • Cadmium-free pigments
  • Environmentally friendly packaging system

Colour: Yellow (2 - 200 µl), blue (50 - 1000 µl)

Length: 50 mm (2 - 200 µl), 70 mm (50 - 1000 µl)

Specifications
  • Colour code:
    Yellow
  • Packed:
    Bulk
  • 容量:
    2 - 200 µl
  • 長さ:
    50 mm
  • Tip colour:
    Yellow
Accessories

Product Information

  • Catalog #
    説明
    使用対象
    Availability
    Price
  • 613-2288
    Tip-Box N, PP, with push on lid, stackable and autoclavable, for 1000 µl tips
    50 - 1000 µl tips
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    使用対象
    Availability
    Price
  • 613-2287
    Tip-Box N, PP, with push on lid, stackable and autoclavable, for 200 µl tips
    2 - 200 µl tips
    Log in for availability
    1 * 1 items

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.