Orders LinkContactUsLinkComponent
VWR®, スプレッダー、L字型、ディスポーザブル
Spreaders, L-shaped, Blue, 5/bag
  612-1561
VWR®, スプレッダー、L字型、ディスポーザブル
  612-1561
VWR®, スプレッダー、L字型、ディスポーザブル
VWR®, スプレッダー、L字型、ディスポーザブル
Spreaders, L-shaped, Blue, 5/bag
1 * 1.000 items
PS, sterile. The foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections. It enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium.
  • Surface treated to enhance adhesion of droplets
  • Non toxic
  • Gamma irradiated
Dimensions: total length 145 mm (±1 mm); spreader length 38 mm (±0,5 mm).
  • Packed:
    5/bag
  • Colour:
    Blue
  • 説明:
    Spreaders, L-shaped