VWR®, スプレッダー、L字型、ディスポーザブル
Spreaders, L-shaped, Blue, 5/bag
PS, sterile. The foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections. It enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium.
- Surface treated to enhance adhesion of droplets
- Non toxic
- Gamma irradiated
Dimensions: total length 145 mm (±1 mm); spreader length 38 mm (±0,5 mm).
- Packed:5/bag
- Colour:Blue
- 説明:Spreaders, L-shaped