VWR®, スプレッダー、L字型、ディスポーザブル
Spreaders, L-shaped, Blue, Individually packed
Supplier: VWR Collection
Jump to:
Product Details & Documents
PS, sterile. The foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections. It enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium.
- Surface treated to enhance adhesion of droplets
- Non toxic
- Gamma irradiated
Dimensions: total length 145 mm (±1 mm); spreader length 38 mm (±0,5 mm).
Documents
Specifications
- Packed:Individually packed
- Colour:Blue
- 説明:Spreaders, L-shaped
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



