VWR®, Calibration Weights, Class M2
200 g, 30 ± mg
Catalog # 611-8361
Supplier: VWR Collection
Jump to:
Product Details & Documents
Test weights in either stainless steel or aluminium.
: Supplied in a plastic box.
Specifications
- Weight:200 g
- Seq:27
- Tolerance:30 ± mg