VWR®, ブロックウェイト、クラスM1
50 kg, 2500 ± mg, With certificate
Catalog # 611-8327
Supplier: VWR Collection
Jump to:
Product Details & Documents
Rectangular weights made from cast iron (density 7,2 g/cm³).
- Indelible unique identification number guarantees traceability
Colour: Grey.
Conforms to OILM, COFRAC accredited
Certifications: Conforms to OIML, COFRAC accredited.
包装: Block weights are supplied with cardboard packaging to protect the weights during transportation.
Documents
Specifications
- Weight:50 kg
- Seq:008
- Tolerance:2500 ± mg
- Type:With certificate