VWR®, ブロックウェイト、クラスM1
10 kg, 500 ± mg, With certificate
Catalog # 611-8325
Supplier: VWR Collection
Jump to:
Product Details & Documents
Rectangular weights made from cast iron (density 7,2 g/cm³).
- Indelible unique identification number guarantees traceability
Colour: Grey.
Conforms to OILM, COFRAC accredited
Certifications: Conforms to OIML, COFRAC accredited.
Packaging: Block weights are supplied with cardboard packaging to protect the weights during transportation.
Specifications
- Weight:10 kg
- Seq:006
- Tolerance:500 ± mg
- Type:With certificate