VWR®, ベンチプロテクタ
Absorption: Approx. 500 ml/m², White, Roll, 460×50 000 mm
Supplier: VWR Collection
Jump to:
Product Details & Documents
High quality, smooth, absorbent paper that quickly absorbs liquid spills with a thick, laminated polyethylene layer that prevents seeping onto the work surface.
- Retains leaked reagents in radiochemical laboratories preventing radioactive contamination
- Enables recovery of expensive leaked materials
- Breakage protection for hard surfaces
- Absorption layer for removing water or solvent from desiccant chambers
- Impermeable protective material for lining animal cages
Absorbance rate: 115-9220 and 115-9221: 500 ml/m²
Absorbance rate: 115-0673: 750 ml/m²
Specifications
- Packed:Roll
- Colour:White
- 説明:Absorption: Approx. 500 ml/m²
- 長さ×幅:460×50 000 mm
- Seq:03



