Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, ベンチプロテクタ

VWR®, ベンチプロテクタ

Absorption: Approx. 500 ml/m², White, Single sheets, 460×570 mm
Catalog # 115-9220
Supplier:   VWR Collection
VWR®, ベンチプロテクタ
Catalog # 115-9220
Supplier:  VWR Collection
VWR®, ベンチプロテクタ
VWR®, ベンチプロテクタ

Absorption: Approx. 500 ml/m², White, Single sheets, 460×570 mm

Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents

High quality, smooth, absorbent paper that quickly absorbs liquid spills with a thick, laminated polyethylene layer that prevents seeping onto the work surface.

  • Retains leaked reagents in radiochemical laboratories preventing radioactive contamination
  • Enables recovery of expensive leaked materials
  • Breakage protection for hard surfaces
  • Absorption layer for removing water or solvent from desiccant chambers
  • Impermeable protective material for lining animal cages

Absorbance rate: 115-9220 and 115-9221: 500 ml/m²
Absorbance rate: 115-0673: 750 ml/m²

Specifications
  • Packed:
    Single sheets
  • Colour:
    White
  • 説明:
    Absorption: Approx. 500 ml/m²
  • 長さ×幅:
    460×570 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.