Orders LinkContactUsLinkComponent
VWR®, ラテックスグローブ
VWR®, ラテックスグローブ
VWR®, ラテックスグローブ
Supplier:  VWR Collection
Certificates
VWR®, ラテックスグローブ
2 Options Available Below
Product Details & Documents
Single-use, ambidextrous gloves made from natural latex.
  • Non sterile, powdered and powder-free
  • Free from phthalates and softeners
  • Textured fingertips for excellent grip
  • Rolled cuffs minimise the risk of tearing
Suitable for food handling, laboratory and industrial applications with complex risks, and for medical applications.
Certifications: EN ISO 374-1 Type C, EN ISO 374-5 VIRUS, EN 455 1-4; ASTM D3578 (except stress at 500% elongation), ASTM F1671; MDR Class I; PPE Cat. III
Food contact approved.
Caution: * Thickness is double measured, minimum values.
Specifications for Product Family
Specifications
  • Catalog No:
  • 112-2764
  • 112-2751
  • Model:
  • Latex
  • Latex
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Finish:
  • Textured fingertips
  • Textured fingertips
  • Cuff style:
  • Beaded
  • Beaded
  • Cuff thickness:
  • 0,12 mm*
  • 0,12 mm*
  • Finger thickness:
  • 0,20 mm*
  • 0,20 mm*
  • Palm thickness:
  • 0,16 mm*
  • 0,16 mm*
  • EN norm:
  • EN ISO 374-1 Type C, EN ISO 374-5 VIRUS, EN 455 1-4
  • EN ISO 374-1 Type C, EN ISO 374-5 VIRUS, EN 455 1-4
  • Food contact approved:
  • +
  • +
Specifications
Product Family Options
Product Information
  • モデル
    手袋のサイズ
    Colour
    長さ
    Packed
    Availability
    Price
  • Powder-free
    XS
    Natural
    240 mm
    100/box
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    112-2764
    Model
    Latex
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
    Finish
    Textured fingertips
    Cuff style
    Beaded
    Cuff thickness
    0,12 mm*
    Finger thickness
    0,20 mm*
    Palm thickness
    0,16 mm*
    EN norm
    EN ISO 374-1 Type C, EN ISO 374-5 VIRUS, EN 455 1-4
    Food contact approved
    +
  • Powder-free
    M
    Natural
    240 mm
    100/box
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    112-2751
    Model
    Latex
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered / powder-free
    Powder-free
    Finish
    Textured fingertips
    Cuff style
    Beaded
    Cuff thickness
    0,12 mm*
    Finger thickness
    0,20 mm*
    Palm thickness
    0,16 mm*
    EN norm
    EN ISO 374-1 Type C, EN ISO 374-5 VIRUS, EN 455 1-4
    Food contact approved
    +