Orders LinkContactUsLinkComponent
VWR®, セルスクレーパーおよびセルリフター
VWR®, セルスクレーパーおよびセルリフター
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, セルスクレーパーおよびセルリフター
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, セルスクレーパーおよびセルリフター
VWR®, セルスクレーパーおよびセルリフター
4 Options Available Below
Product Details & Documents
Specifically designed to make collecting cells easier and more effective. Available with a choice of blade positions: scraper for use in flasks, or lifter for use in harvesting cells (especially stem cells) in dishes.
  • Cross-ribbed handle in ABS provides greater rigidity and ensures better control while scraping cells
  • Thin, flexible TPE blade prevents damage to cells
  • Individually wrapped
  • Sterilised by electron beam irradiation
  • Free from DNase, RNase and non-pyrogenic
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-2602
  • 734-2603
  • 734-2604
  • 734-2605
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Product Information
  • 説明
    総長
    Availability
    Price
  • Cell scraper, blade width 20 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2602
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Cell lifter, blade width 20 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2603
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Cell scraper, blade width 30 mm
    390 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2604
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Cell lifter, blade width 30 mm
    390 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-2605
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M