Orders LinkContactUsLinkComponent
VWR®, フロアマットフレーム、PureStepマット用
VWR®, フロアマットフレーム、PureStepマット用
Supplier:   VWR Collection
VWR®, フロアマットフレーム、PureStepマット用
Supplier:  VWR Collection
Certificates
VWR®, フロアマットフレーム、PureStepマット用
VWR®, フロアマットフレーム、PureStepマット用
8 Options Available Below
Product Details & Documents
These frames are designed to be used with PureStep mats to create a system, that is ideal for any critical area, and does not allow direct adhesion to the flooring surface. The frames are constructed of a hard, rigid, polystyrene base material, combined with a non skid backing, that holds the mats in place on almost any surface.
  • Protects floors from adhesive residue
  • Reduces the risk of slips and falls
  • Non slip parts made from polymer coated, polyester textile
  • Naturally antimicrobial, mould and mildew resistant
注文情報: Adhesive mats are sold separately.
Product Family Options
Product Information
  • 説明
    Colour
    For
    長さ×幅
    厚さ
    Availability
    Price
  • Mat frame
    White
    457×914 mm
    508×965 mm
    3,2 mm
    1 * 1 items
  • Mat frame
    White
    457×1143 mm
    508×1194 mm
    3,2 mm
    1 * 1 items
  • Mat frame
    White
    609×914 mm
    660×965 mm
    3,2 mm
    1 * 1 items
  • Mat frame for 609.6×762 mm mats, white
    635×787,4 mm
    1 * 1 items
  • Mat frame
    White
    660×1143 mm
    711×1193 mm
    3,2 mm
    1 * 1 items
  • Mat frame
    White
    914×914 mm
    965×965 mm
    3,2 mm
    1 * 1 items
  • Mat frame
    White
    914×1140 mm
    965×1194 mm
    3,2
    1 * 1 items
  • Mat frame
    White
    914×1524 mm
    965×1574 mm
    3,2 mm