Orders LinkContactUsLinkComponent
VWR®, ラベリングテープ、ウェハーボックスシール
VWR®, ラベリングテープ、ウェハーボックスシール
Supplier:   VWR Collection
VWR®, ラベリングテープ、ウェハーボックスシール
Supplier:  VWR Collection
Certificates
VWR®, ラベリングテープ、ウェハーボックスシール
VWR®, ラベリングテープ、ウェハーボックスシール
6 Options Available Below
Product Details & Documents
These tapes, with polyethylene backing and acrylic adhesive, have low out-gassing and good clean-peel properties. They are mostly used for colour coding and wafer box sealing.
  • Over labelling (to protect paper labels)
  • Maintenance
  • Colour coding
Processed in a controlled environment, tapes are wound on a plastic core and single-bagged.
Length: 32,9 m
Different colours available.
Product Family Options
Product Information
  • 説明
    Colour
    長さ
    Availability
    Price
  • Wafer box sealing cleanroom tape
    White
    32,9 m
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll
  • Wafer box sealing cleanroom tape
    Dark blue
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 4 Roll
    1 * 4 Roll
  • Wafer box sealing cleanroom tape, metal-free
    Transparent
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 36 Roll
    1 * 36 Roll
  • Wafer box sealing cleanroom tape
    Transparent
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 36 Roll
    1 * 36 Roll
  • Wafer box sealing cleanroom tape
    White
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 24 Roll
    1 * 24 Roll
  • Wafer box sealing cleanroom tape
    Transparent
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 24 Roll
    1 * 24 Roll