Orders LinkContactUsLinkComponent
VWR®, 試験管ラック、PP
VWR®, 試験管ラック、PP
VWR®, 試験管ラック、PP
Supplier:  VWR Collection
Certificates
VWR®, 試験管ラック、PP
VWR®, 試験管ラック、PP
8 Options Available Below
Product Details & Documents
PP.
  • Single-piece moulded
  • Good heat and chemical resistance
  • Three tiers with contents always clearly visible
  • Large labelling areas on the side surfaces
Product Family Options
Product Information
  • Colour
    穴の数
    For tube Ø
    W×D×H
    Availability
    Price
  • Blue
    72 (6×12)
    13 mm
    103,5×202×60 mm
    1 * 1 items
  • Blue
    72 (6×12)
    16 mm
    127,3×250,6×73 mm
    1 * 1 items
  • Green
    72 (6×12)
    13 mm
    103,5×202×60 mm
    1 * 1 items
  • Green
    72 (6×12)
    16 mm
    127,3×250,6×73 mm
    1 * 1 items
  • Magenta
    72 (6×12)
    13 mm
    103,5×202×60 mm
    1 * 1 items
  • Red
    72 (6×12)
    16 mm
    127,3×250,6×73 mm
    1 * 1 items
  • White
    72 (6×12)
    13 mm
    103,5×202×60 mm
    1 * 1 items
  • White
    72 (6×12)
    16 mm
    127,3×250,6×73 mm