Orders LinkContactUsLinkComponent
VWR®, 漏斗、分析用
VWR®, 漏斗、分析用
VWR®, 漏斗、分析用
Supplier:  VWR Collection
Certificates
VWR®, 漏斗、分析用
VWR®, 漏斗、分析用
16 Options Available Below
Product Details & Documents
PP, translucent.
  • Inner ribs at an angle of 58° for rapid filtration
  • Outer ribs prevent air from being trapped under the filter paper
  • Very good chemical and temperature resistance
Designed for use with standard filter paper. Cone angle 60°.
Product Family Options
Product Information
  • 長さ
    Stem length
    Stem O-Ø
    Funnel O-Ø
    Availability
    Price
  • 75,8 mm
    49,3 mm
    6,3 mm
    35 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items
  • 75,8 mm
    49,3 mm
    6,3 mm
    35 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - $53.35
    $53.35
    1 * 12 items
       $53.35
  • 84,6 mm
    52,0 mm
    7,1 mm
    45 mm
    Log in for availability
    Log in for availability
  • 84,6 mm
    52,0 mm
    7,1 mm
    45 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - $57.37
    $57.37
    1 * 12 items
       $57.37
  • 103,4 mm
    59,5 mm
    7,1 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items
  • 103,4 mm
    59,5 mm
    7,1 mm
    55 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - $60.77
    $60.77
    1 * 12 items
       $60.77
  • 111,7 mm
    61,4 mm
    6,4 mm
    65 mm
    Log in for availability
    Log in for availability
  • 111,7 mm
    61,4 mm
    6,4 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
  • 111,7 mm
    61,4 mm
    6,4 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 12 items
    1 * 12 items - $69.02
    $69.02
    1 * 12 items
       $69.02
  • 139,4 mm
    76,3 mm
    6,8 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items
  • 139,4 mm
    76,3 mm
    6,8 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - $48.78
    $48.78
    1 * 6 items
       $48.78
  • 164,0 mm
    89,0 mm
    8,2 mm
    90 mm
    Log in for availability
    Log in for availability
  • 164,0 mm
    89,0 mm
    8,2 mm
    90 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - $57.52
    $57.52
    1 * 6 items
       $57.52
  • 180,0 mm
    95,4 mm
    9,1 mm
    100 mm
    Log in for availability
    Log in for availability
  • 180,0 mm
    95,4 mm
    9,1 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items - $46.76
    $46.76
    1 * 4 items
       $46.76
  • 280,0 mm
    154,0 mm
    14,0 mm
    160 mm