Ethidium bromide 0,625 mg/l in aqueous solution, Biotechnology Grade
Catalog # E406-5ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- パックタイプ:Dropping bottle
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Abs.@ Lambda Max:0.68 – 0.74
- DNase:none detected
- Lambda Max (8/100 ml Water):478 – 482 nm
- RNase:none detected
Specification Test Results
About this item
:
:394.31 g/mol
EINECS:214-984-6
CAS Number:1239-45-8
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724