Orders LinkContactUsLinkComponent
SASスーパー、VWR®用アクセサリー
SASスーパー、VWR®用アクセサリー
SASスーパー、VWR®用アクセサリー
Supplier:  VWR Collection
Certificates
SASスーパー、VWR®用アクセサリー
SASスーパー、VWR®用アクセサリー
21 Options Available Below
Product Family Options
Product Information
  • 説明
    Availability
    Price
  • Adapter for Petri dishes Ø 90 mm
    1 * 1 items
  • Aluminum carrying case
    1 * 1 items
  • Battery charger with interchangeable plug, 90 – 260 V
    1 * 1 items
  • Downloading device for software
    1 * 1 items
  • Infrared remote control
    1 * 1 items
  • IQ-OQ-PQ manual
    1 * 1 items
  • IQ-OQ-PQ validation protocols
    1 * 1 items
  • IQ-OQ-PQ validation protocols
    1 * 1 items
  • Lightweight aluminium carrying case
    1 * 1 items
  • Soft carryng case for SAS 100/180 air samplers
    1 * 1 items
  • Soft carrying case for SAS Super DUO
    1 * 1 items
  • Software for downloading data
    1 * 1 items
  • Stainless steel holder for table and wall
    1 * 1 items
  • Telescopic floor tripod, extendable up to 2800 mm high
    1 * 1 items
  • Plastic cover for SAS head, autoclavable
    1 * 1 items
Heads for contact plates (55 mm)
Product Information
  • 説明
    Availability
    Price
  • Aluminum aspirating head for Ø 55 mm contact plates
    1 * 1 items
  • Stainless steel aspirating head for Ø 55 mm contact plates
    1 * 1 items
Heads for Petri dishes (90 mm)
Product Information
  • 説明
    Availability
    Price
  • Aluminium head and adapter for standard Ø 90 mm Petri dishes
    1 * 1 items
  • Aluminium head for standard Ø 90 mm Petri dishes (without adapter)
    1 * 1 items
  • Stainless steel head and adapter for standard Ø 90 mm Petri dishes
    1 * 1 items
  • Stainless steel head for standard Ø 90 mm Petri dishes (without adapter)