Orders LinkContactUsLinkComponent
VWR®, シューズカバー、CPE
VWR®, シューズカバー、CPE
VWR®, シューズカバー、CPE
Supplier:  VWR Collection
Certificates
VWR®, シューズカバー、CPE
3 Options Available Below
Product Details & Documents
Basic shoe covers manufactured from a low-density chlorinated polyethylene fabric, which is designed to protect against dirt, grime, and liquid splash.
  • CPE is water resistant and provides good resistance against chemicals
  • Economical and lint-free
Ideal for laboratory use.
* One size fits all
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-1491
  • VWRU414004-513
  • 113-1492
  • Garment application:
  • Laboratory
  • Laboratory
  • Laboratory
Specifications
Product Family Options
Product Information
  • Colour
    靴のサイズ
    Availability
    Price
  • White
    U*
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-1491
    Garment application
    Laboratory
  • White
    U*
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    VWRU414004-513
    Garment application
    Laboratory
  • Blue
    U*
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-1492
    Garment application
    Laboratory