Orders LinkContactUsLinkComponent
保存システム、Cryoinstant
保存システム、Cryoinstant
保存システム、Cryoinstant
Supplier:  VWR Chemicals
Certificates
保存システム、Cryoinstant
7 Options Available Below
Product Details & Documents
Microbiological laboratories need a simple procedure to maintain important microorganisms rather than repetitive sub-culturing, which results in contaminated cultures, loss of viability and, even more importantly, loss of original characteristics by introducing biochemical and genetic changes. Cryoinstant consists of 25 porous beads in a vial filled with a broth containing glycerol. Microorganisms will bind to the porous surface of the beads. The excess broth is aspirated, and the vials stored at temperatures down to –70 °C in a freezer.
  • Quick and easy to use
  • Colour coded
  • Storage down to –70 °C
  • Quality controlled for fertility and sterility
Packaging: Cryoinstant is packed in packs of 50 vials of 2 ml.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 822075ZA
  • 822070ZA
  • 822071ZA
  • 822072ZA
  • 822073ZA
  • 822074ZA
  • 820006ZA
  • Shelf life:
  • 1440 days
  • 1440
  • 1440
  • 1440
  • 1440
  • 1440
  • 1440
  • Storage temperature:
  • 8...25 °C
Specifications
Product Family Options
Product Information
  • 説明
    Pk
    Availability
    Price
  • Cryoinstant, natural
    50
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    822075ZA
    Shelf life
    1440 days
    Storage temperature
  • Cryoinstant, mixed
    50
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    822070ZA
    Shelf life
    1440
    Storage temperature
  • Cryoinstant, red
    50
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    822071ZA
    Shelf life
    1440
    Storage temperature
  • Cryoinstant, blue
    50
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    822072ZA
    Shelf life
    1440
    Storage temperature
  • Cryoinstant, green
    50
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    822073ZA
    Shelf life
    1440
    Storage temperature
  • Cryoinstant, yellow
    50
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    822074ZA
    Shelf life
    1440
    Storage temperature
  • Cryoinstant, vials without media
    50
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    820006ZA
    Shelf life
    1440
    Storage temperature
    8...25 °C