Orders LinkContactUsLinkComponent
VWR®, スライドメーラー
VWR®, スライドメーラー
VWR®, スライドメーラー
VWR®, スライドメーラー
These boxes are designed for shipping, storing, or staining slides.
  • Inside wall features slotted channels to keep slides separated and making removal easier
  • Closes securely to keep slides safe - side opening
  • Assorted colours (red, yellow, blue, purple, orange) for easier identification
Product Family Options
  • 説明
    Colour
    Material
    W×D×H
    No. of slides
  • Slide mailers, with end opening
    Assorted
    PP
    43×24×88 mm
    5
    1 * 25 items
    1 * 25 items - $34.48
    $34.48
    1 * 25 items
       $34.48
  • Slide mailers, with side opening
    Natural
    PP
    81×19×31 mm
    5
    1 * 25 items
    1 * 25 items