VWR®, ピペットラック
Cargo manual pipette racks, Blue, 95×127,5×301 mm
612-3848
Ideal for serological pipettes.
- Click together horizontally with side connections
- Securely lock in place with top connections
- Magnets on both sides of units connect securely and safely to metal surface, such as filling cabinets, fume hoods, refrigerators and freezers
- Angled shelves store 0,1 to 50 ml pipettes within arm's reach
- Protective lid encloses the unit to prevent dust build up, and provides an additional shelf for extra space when connected horizontally
- Colour:Blue
- 説明:Cargo manual pipette racks
- W×D×H:95×127,5×301 mm