Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Japan
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ラック
Tube Racks
VWR®, Test Tube Racks, Fold and Snap
VWR®, Test Tube Racks, Fold and Snap
:
VWR Collection
VWR®, Test Tube Racks, Fold and Snap
:
VWR Collection
PP.
Easy to assemble polypropylene racks snap together in seconds
Designed to remain stable in water baths
Shipped flat
CoC VWR PP Test Tube Racks, Fold and Snap
COC_VWRI211-0353_516556
COC_VWRI211-0359_518749
VWR® Collection Selection Catalogue
VWR® Lab bric-a-brac
Frequently Bought Together
VWR®, Test Tube Racks, Fold and Snap
: VWR Collection