Orders LinkContactUsLinkComponent
VWR®, 滴下ボトル、PTFE
VWR®, 滴下ボトル、PTFE
Supplier:   VWR Collection
VWR®, 滴下ボトル、PTFE
Supplier:  VWR Collection
Certificates
VWR®, 滴下ボトル、PTFE
VWR®, 滴下ボトル、PTFE
2 Options Available Below
Product Details & Documents
PTFE, white.
  • Flexible wall for easy use
  • Screw-fit cap
  • Guaranteed chemically resistant and inert
Suitable for dropping-out aggressive chemicals.
Product Family Options
Product Information
  • 容量
    高さ
    外径
    Availability
    Price
  • 25 ml
    80 mm
    33 mm
    1 * 1 items
  • 50 ml
    100 mm
    43 mm