Orders LinkContactUsLinkComponent
VWR® Mega, 試験管ラック
Supplier:   VWR Collection
VWR® Mega, 試験管ラック
Supplier:  VWR Collection
Certificates
VWR® Mega, 試験管ラック
VWR® Mega, 試験管ラック
8 Options Available Below
Product Details & Documents
High capacity storage for your tubes.
  • Sturdy, material for easy and secure transport of blood collection and other tubes
  • Alphanumeric identification for quick location and retrieval of tubes
  • Place different test groupings into one rack eliminating the need for multiple racks
  • Autoclavable
Fully assembled, polypropylene mega racks are great for high throughput labs requiring large sample storage.
Product Family Options
Product Information
  • 説明
    穴の直径
    Colour
    穴の数
    For tubes
    W×D×H
    Availability
    Price
  • Single size
    16 mm
    White
    120
    10 ml
    337×267×91 mm
    1 * 1 items
  • Single size
    16 mm
    Blue
    120
    10 ml
    337×267×91 mm
    1 * 1 items
  • Double size
    16 mm
    Blue
    240
    10 ml
    673×267×90 mm
    1 * 1 items
  • Double size
    16 mm
    White
    240
    10 ml
    673×267×90 mm
    1 * 1 items
  • Single size
    13 mm
    White
    216
    5 - 7 ml
    337×325×91 mm
    1 * 1 items
  • Single size
    13 mm
    Blue
    216
    5 - 7 ml
    337×325×91 mm
    1 * 1 items
  • Double size
    13 mm
    White
    432
    5 - 7 ml
    673×235×90 mm
    1 * 1 items
  • Double size
    13 mm
    Blue
    432
    5 - 7 ml
    673×235×90 mm