Orders LinkContactUsLinkComponent
VWR®, Polyester Films with Colour-Coded Marking Areas
Supplier:   VWR Collection
VWR®, Polyester Films with Colour-Coded Marking Areas
Supplier:  VWR Collection
Certificates
VWR®, Polyester Films with Colour-Coded Marking Areas
VWR®, Polyester Films with Colour-Coded Marking Areas
5 Options Available Below
Product Details & Documents
Identical to polyester films above, but include a colour dot and white writing area on each end tab for plate identification. End tabs with backing removed adhere to the ends of the plate and are perforated so they can be left in place even if the centre portion of the film is removed. Alternatively, one inscribed colour-coded end tab can be removed and pasted in a laboratory notebook while the other remains on the plate.
  • Choice of five colours for easy plate identification
  • Applications: Short-term storage, incubation and ELISA
  • Recommended temperature range −40...+120 °C
Product Family Options
Product Information
  • 説明
    光学特性
    厚さ
    Sterile
    Availability
    Price
  • Films with purple dot
    Transparent
    77,5 µm
    -
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    391-1267
  • Films with orange dot
    Transparent
    77,5 µm
    -
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    391-1268
  • Films with red dot
    Transparent
    77,5 µm
    -
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    391-1269
  • Films with blue dot
    Transparent
    77,5 µm
    -
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    391-1270
  • Films with assorted dots
    Transparent
    77,5 µm
    -
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    391-1271