Orders LinkContactUsLinkComponent
VWR®, ポリエステル2×8プレートストリップフィルム
VWR®, ポリエステル2×8プレートストリップフィルム
Supplier:   VWR Collection
VWR®, ポリエステル2×8プレートストリップフィルム
Supplier:  VWR Collection
Certificates
VWR®, ポリエステル2×8プレートストリップフィルム
VWR®, ポリエステル2×8プレートストリップフィルム
2 Options Available Below
Product Details & Documents
Polyester 2×8 strip films are identical to polyester films in material and adhesive, but designed for sealing only one or two 8-well rows at a time on either strip-well plates or standard plates whenever rows must be selectively protected or accessed. Sterile product is packed in tamper-evident bags of 50/bag.
  • Seal selectively, one or two rows at a time
  • Can use as secondary seal over previously pierced films
  • Recommended temperature range −40 to +120 °C
Each film L×D: 103,2×19,1 mm, including two removable positioning tabs; long dimension with tabs removed is 82,6 mm.
Product Family Options
Product Information
  • 説明
    光学特性
    厚さ
    Sterile
    Availability
    Price
  • Polyester 2×8 films
    Transparent
    77,5 µm
    -
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items
  • Polyester 2×8 films
    Transparent
    77,5 µm
    +
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items