Orders LinkContactUsLinkComponent
レフィル用セット, SPILL-X®
レフィル用セット, SPILL-X®
レフィル用セット, SPILL-X®
レフィル用セット, SPILL-X®
These refill sets can be used with five types of SPILL-X® treatment kits. They are available in different formats.
  • Each set contains six bottles
 : Multipurpose SPILL-X® set contains 2 acids, 2 caustics and 2 solvent agents, whereas the formaldehyde SPILL-X-FP® refill set comprises 6 bottles of formaldehyde agents.
Product Family Options
  • 説明
  • Acid refill, SPILL-X®, 6 bottle set
    1 * 1 SET
    1 * 1 SET
  • Multipurpose refill, SPILL-X®
    1 * 1 SET
    1 * 1 SET
  • Solvent refill, SPILL-X®, 6 bottle set
    1 * 1 SET
    1 * 1 SET
  • Formaldehyde refill, SPILL-X-FP®, 6 bottle set
    1 * 1 SET
    1 * 1 SET