VWR®, 化学物質用流出対応キット
Spill response kit for chemicals, 10 l/kit
Catalog # 121-1050
Supplier: VWR Collection
Jump to:
Product Details & Documents
Spill kit suitable for dangerous liquids.
- Total absorbent capacity up to 10 litres, depending on the chemical being absorbed
- Suitable for oils and solvents
- Can be used for most chemicals
Supplied in a clip closed clear bag.
Delivery information: Kit contains: 2× blue plastic disposal bags, 1× 6 cm×1,2m maintenance sock, 1× clip close carrier, 1× instruction sheet, 10× maintenance spill pads, 1× non branded spill kit header card, 2× plastic zip ties.
Documents
121-1050
Images 360
TDS_VWR_SPILL_RESPONSE_KIT_FOR_CHEMICALS.pdf
Product specification
VWR® Lab bric-a-brac
Product Literature
Specifications
- 説明:Spill response kit for chemicals
- Absorbency:10 l/kit