Orders LinkContactUsLinkComponent
VWR®, 防振秤量面
Supplier:   VWR Collection
VWR®, 防振秤量面
Supplier:  VWR Collection
Certificates
VWR®, 防振秤量面
VWR®, 防振秤量面
2 Options Available Below
Product Details & Documents
Polished granite. Four absorbing feet dampen the transmission of extraneous vibrations to this slab making it suitable, not only for high precision weighing, but also microscopy and other similar delicate instrumentation.
Weight: 44 kg
Product Family Options
Product Information
  • 説明
    W×D×H
    Weight
    Availability
    Price
  • Anti-vibration weighing surface, granite
    560×450×65 mm
    51 kg
    1 * 1 items
  • Anti-vibration weighing surface
    450x300x40 mm
    20 kg