VWR®, フィルターチップ
Bevel Point, graduated, 1 - 100 µl, +, +, 49,66 mm, 10 racks, 96 each
Supplier: VWR Collection
VWR®, フィルターチップ
Catalog # 732-1102
Supplier: VWR Collection
Jump to:
Product Details & Documents
All tips are made of high quality, 100% pure, virgin medical grade PP. Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard. All products are tested for endotoxin (pyrogen) contamination and certified to USP results. Resins are pre-tested for metal contamination. Certificates are on file by lot number for all test results.
- A wider seal ensures that tips will seal easily and consistently without leaking, on old as well as new pipettes
- Guaranteed free of detectable human DNA, DNase/RNase, ATP, endotoxins and heavy metals
Specifications
- CE-IVD:+
- Packed:10 racks, 96 each
- Sterile:+
- 容量:1 - 100 µl
- 長さ:49,66 mm
- Seq:34
- Tip style:Bevel Point, graduated
- Cat. No.:732-1102
- Environmentally Preferable:Low Manufacturing Impact|Sustainable Packaging