VWR®, 定量用フィルターペーパー
About this item
424: Filtration of lead sulphide, iron sulphide, silver sulphide, alkali carbonate, lead chromate, arsenic, antimony, cadmium and foodstuffs. Determination of sand content in dairy products. Determination of whiteness of pulp in paper mills.
434: Analysis of very fine precipitates such as lead dioxide, calcium fluoride, nickel sulphide, zinc sulphide, cold barium sulphate and cuprous oxide. Determination of insoluble impurities in greases, animal and vegetable oils.
454: Retention of zirconium hydroxide, aluminium hydroxide, cobalt sulphide and other coarse or gelatinous precipitates.
474: General quantitative analyses. Also for filtration of aluminium phosphate, zinc ammonium phosphate and silver chromate.
23 Options Available Below
Product Details & Documents
424: Filtration of lead sulphide, iron sulphide, silver sulphide, alkali carbonate, lead chromate, arsenic, antimony, cadmium and foodstuffs. Determination of sand content in dairy products. Determination of whiteness of pulp in paper mills.
434: Analysis of very fine precipitates such as lead dioxide, calcium fluoride, nickel sulphide, zinc sulphide, cold barium sulphate and cuprous oxide. Determination of insoluble impurities in greases, animal and vegetable oils.
454: Retention of zirconium hydroxide, aluminium hydroxide, cobalt sulphide and other coarse or gelatinous precipitates.
474: General quantitative analyses. Also for filtration of aluminium phosphate, zinc ammonium phosphate and silver chromate.
Specifications for Product Family
Specifications
- Catalog No:
- 516-0264
- 516-0265
- 516-0266
- 516-0267
- 516-0268
- 516-0269
- 516-0270
- 516-0271
- 516-0272
- 516-0273
- 516-0274
- 516-0275
- 516-0847
- 516-0848
- 516-0849
- 516-0850
- 516-0263
- 516-0853
- 516-0854
- 516-0855
- 516-0856
- 516-0857
- VWRU28306-380
- Grade:
- 424
- 424
- 424
- 424
- 424
- 424
- 434
- 434
- 434
- 434
- 434
- 434
- 474
- 474
- 474
- 474
- 474
- 454
- 454
- 454
- 454
- 454
- 474
- Weight:
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- 84 g/m²
- Thickness:
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,17 mm
- 0,14 mm
- 0,14 mm
- 0,14 mm
- 0,14 mm
- 0,14 mm
- 0,14 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,16 mm
- 0,18 mm
- 0,18 mm
- 0,18 mm
- 0,18 mm
- 0,18 mm
- 0,16 mm
- Particle retention:
- 8 - 12 µm
- 8 - 12 µm
- 8 - 12 µm
- 8 - 12 µm
- 8 - 12 µm
- 8 - 12 µm
- 2 - 3 µm
- 2 - 3 µm
- 2 - 3 µm
- 2 - 3 µm
- 2 - 3 µm
- 2 - 3 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- 5 - 8 µm
- 15 - 12 µm
- 15 - 12 µm
- 15 - 12 µm
- 15 - 12 µm
- 15 - 12 µm
- 5 - 8 µm
- Max. pore size:
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- 40 µm
- 20 µm
- 20 µm
- 20 µm
- 20 µm
- 20 µm
- 20 µm
- 30 µm
- 30 µm
- 30 µm
- 30 µm
- 30 µm
- 50 µm
- 50 µm
- 50 µm
- 50 µm
- 50 µm
- 30 µm
- Min. pore size:
- 23 µm
- 23 µm
- 23 µm
- 23 µm
- 23 µm
- 23 µm
- 9 µm
- 9 µm
- 9 µm
- 9 µm
- 9 µm
- 9 µm
- 15 µm
- 15 µm
- 15 µm
- 15 µm
- 15 µm
- 31 µm
- 31 µm
- 31 µm
- 31 µm
- 31 µm
- 15 µm
- Filtration speed:
- 150 s (Herzberg)
- 150 s (Herzberg)
- 150 s (Herzberg)
- 150 s (Herzberg)
- 150 s (Herzberg)
- 150 s (Herzberg)
- 2500 s (Herzberg)
- 2500 s (Herzberg)
- 2500 s (Herzberg)
- 2500 s (Herzberg)
- 2500 s (Herzberg)
- 2500 s (Herzberg)
- 450 s (Herzberg)
- 450 s (Herzberg)
- 450 s (Herzberg)
- 450 s (Herzberg)
- 450 s (Herzberg)
- 70 s (Herzberg)
- 70 s (Herzberg)
- 70 s (Herzberg)
- 70 s (Herzberg)
- 70 s (Herzberg)
- 450 s (Herzberg)
- Wet burst strength:
- 2,5 psi
- 2,5 psi
- 2,5 psi
- 2,5 psi
- 2,5 psi
- 2,5 psi
- 3,5 psi
- 3,5 psi
- 3,5 psi
- 3,5 psi
- 3,5 psi
- 3,5 psi
- 3 psi
- 3 psi
- 3 psi
- 3 psi
- 3 psi
- 2 psi
- 2 psi
- 2 psi
- 2 psi
- 2 psi
- 3 psi
- Dry burst strength:
- 17 psi
- 17 psi
- 17 psi
- 17 psi
- 17 psi
- 17 psi
- 18 psi
- 18 psi
- 18 psi
- 18 psi
- 18 psi
- 18 psi
- 12 psi
- 12 psi
- 12 psi
- 12 psi
- 12 psi
- 11 psi
- 11 psi
- 11 psi
- 11 psi
- 11 psi
- 12 psi
Specifications
Product Family Options
Product Information
- GradeサイズAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.516-0264Grade424Weight84 g/m²Thickness0,17 mmParticle retention8 - 12 µmMax. pore size40 µmMin. pore size23 µmFiltration speed150 s (Herzberg)Wet burst strength2,5 psiDry burst strength17 psi - Specifications:
More
Specifications
Cat. No.516-0265Grade424Weight84 g/m²Thickness0,17 mmParticle retention8 - 12 µmMax. pore size40 µmMin. pore size23 µmFiltration speed150 s (Herzberg)Wet burst strength2,5 psiDry burst strength17 psi - Specifications:
More
Specifications
Cat. No.516-0266Grade424Weight84 g/m²Thickness0,17 mmParticle retention8 - 12 µmMax. pore size40 µmMin. pore size23 µmFiltration speed150 s (Herzberg)Wet burst strength2,5 psiDry burst strength17 psi - Specifications:
More
Specifications
Cat. No.516-0267Grade424Weight84 g/m²Thickness0,17 mmParticle retention8 - 12 µmMax. pore size40 µmMin. pore size23 µmFiltration speed150 s (Herzberg)Wet burst strength2,5 psiDry burst strength17 psi - Specifications:
More
Specifications
Cat. No.516-0268Grade424Weight84 g/m²Thickness0,17 mmParticle retention8 - 12 µmMax. pore size40 µmMin. pore size23 µmFiltration speed150 s (Herzberg)Wet burst strength2,5 psiDry burst strength17 psi - Specifications:
More
Specifications
Cat. No.516-0269Grade424Weight84 g/m²Thickness0,17 mmParticle retention8 - 12 µmMax. pore size40 µmMin. pore size23 µmFiltration speed150 s (Herzberg)Wet burst strength2,5 psiDry burst strength17 psi - Specifications:
More
Specifications
Cat. No.516-0270Grade434Weight84 g/m²Thickness0,14 mmParticle retention2 - 3 µmMax. pore size20 µmMin. pore size9 µmFiltration speed2500 s (Herzberg)Wet burst strength3,5 psiDry burst strength18 psi - Specifications:
More
Specifications
Cat. No.516-0271Grade434Weight84 g/m²Thickness0,14 mmParticle retention2 - 3 µmMax. pore size20 µmMin. pore size9 µmFiltration speed2500 s (Herzberg)Wet burst strength3,5 psiDry burst strength18 psi - Specifications:
More
Specifications
Cat. No.516-0272Grade434Weight84 g/m²Thickness0,14 mmParticle retention2 - 3 µmMax. pore size20 µmMin. pore size9 µmFiltration speed2500 s (Herzberg)Wet burst strength3,5 psiDry burst strength18 psi - Specifications:
More
Specifications
Cat. No.516-0273Grade434Weight84 g/m²Thickness0,14 mmParticle retention2 - 3 µmMax. pore size20 µmMin. pore size9 µmFiltration speed2500 s (Herzberg)Wet burst strength3,5 psiDry burst strength18 psi - Specifications:
More
Specifications
Cat. No.516-0274Grade434Weight84 g/m²Thickness0,14 mmParticle retention2 - 3 µmMax. pore size20 µmMin. pore size9 µmFiltration speed2500 s (Herzberg)Wet burst strength3,5 psiDry burst strength18 psi - Specifications:
More
Specifications
Cat. No.516-0275Grade434Weight84 g/m²Thickness0,14 mmParticle retention2 - 3 µmMax. pore size20 µmMin. pore size9 µmFiltration speed2500 s (Herzberg)Wet burst strength3,5 psiDry burst strength18 psi - Specifications:
More
Specifications
Cat. No.516-0847Grade474Weight84 g/m²Thickness0,16 mmParticle retention5 - 8 µmMax. pore size30 µmMin. pore size15 µmFiltration speed450 s (Herzberg)Wet burst strength3 psiDry burst strength12 psi - Specifications:
More
Specifications
Cat. No.516-0848Grade474Weight84 g/m²Thickness0,16 mmParticle retention5 - 8 µmMax. pore size30 µmMin. pore size15 µmFiltration speed450 s (Herzberg)Wet burst strength3 psiDry burst strength12 psi - Specifications:
More
Specifications
Cat. No.516-0849Grade474Weight84 g/m²Thickness0,16 mmParticle retention5 - 8 µmMax. pore size30 µmMin. pore size15 µmFiltration speed450 s (Herzberg)Wet burst strength3 psiDry burst strength12 psi - Specifications:
More
Specifications
Cat. No.516-0850Grade474Weight84 g/m²Thickness0,16 mmParticle retention5 - 8 µmMax. pore size30 µmMin. pore size15 µmFiltration speed450 s (Herzberg)Wet burst strength3 psiDry burst strength12 psi - Specifications:
More
Specifications
Cat. No.516-0263Grade474Weight84 g/m²Thickness0,16 mmParticle retention5 - 8 µmMax. pore size30 µmMin. pore size15 µmFiltration speed450 s (Herzberg)Wet burst strength3 psiDry burst strength12 psi - Specifications:
More
Specifications
Cat. No.516-0853Grade454Weight84 g/m²Thickness0,18 mmParticle retention15 - 12 µmMax. pore size50 µmMin. pore size31 µmFiltration speed70 s (Herzberg)Wet burst strength2 psiDry burst strength11 psi - Specifications:
More
Specifications
Cat. No.516-0854Grade454Weight84 g/m²Thickness0,18 mmParticle retention15 - 12 µmMax. pore size50 µmMin. pore size31 µmFiltration speed70 s (Herzberg)Wet burst strength2 psiDry burst strength11 psi - Specifications:
More
Specifications
Cat. No.516-0855Grade454Weight84 g/m²Thickness0,18 mmParticle retention15 - 12 µmMax. pore size50 µmMin. pore size31 µmFiltration speed70 s (Herzberg)Wet burst strength2 psiDry burst strength11 psi - Specifications:
More
Specifications
Cat. No.516-0856Grade454Weight84 g/m²Thickness0,18 mmParticle retention15 - 12 µmMax. pore size50 µmMin. pore size31 µmFiltration speed70 s (Herzberg)Wet burst strength2 psiDry burst strength11 psi - Specifications:
More
Specifications
Cat. No.516-0857Grade454Weight84 g/m²Thickness0,18 mmParticle retention15 - 12 µmMax. pore size50 µmMin. pore size31 µmFiltration speed70 s (Herzberg)Wet burst strength2 psiDry burst strength11 psi - Specifications:
More
Specifications
Cat. No.VWRU28306-380Grade474Weight84 g/m²Thickness0,16 mmParticle retention5 - 8 µmMax. pore size30 µmMin. pore size15 µmFiltration speed450 s (Herzberg)Wet burst strength3 psiDry burst strength12 psi