吸水体ベンチ下敷き、VWR® (英語版のみ
Deluxe bench underpads, Blue, 914×584 mm, 680 ml
Catalog # 115-0683
Supplier: VWR Collection
Jump to:
Product Details & Documents
Absorbent underpads with a three-layer construction made from PP top and back sheets and a cellulose filler.
- Top sheet made from white spunbound polypropylene
- Absorbent cellulose fluff filler between two layers of tissue for superior wicking of fluid
- Cushion effect to protect labware from breakage
- Embossed PP backing sheet comes in different colours
- Sealed on four sides to prevent liquid seepage
Deluxe pads are more absorbent and thicker with more cellulose fluff than standard pads.
: Super Deluxe pads have an added polymer that absorbs more moisture.
Specifications
- Colour:Blue
- 説明:Deluxe bench underpads
- 長さ×幅:914×584 mm
- Seq:12
- Absorbency:680 ml