VWR®, 使い捨て計量ボート
90×90×7 mm
Catalog # 611-2249
Supplier: VWR Collection
Jump to:
Product Details & Documents
These flexible PS boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
- Shallow boat with a wide, flat bottom to resist tipping
- Temperature range –10 to 70 °C (14 to 158 °F)
- Can be used with microwave moisture analysers
- All are FDA/food-grade compliant
- Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases
These sturdy weighing boats are ideal for weighing solid products.
Documents
CoC VWR Disposable Weighing Boats.pdf
Conformity declaration
VWR® Collection Selection Catalogue VWR® for Weighing
Product Literature
Specifications
- W×D×H:90×90×7 mm