Orders LinkContactUsLinkComponent
メスシリンダー、トールフォーム、PP
メスシリンダー、トールフォーム、PP
Supplier:   VWR Collection
メスシリンダー、トールフォーム、PP
Supplier:  VWR Collection
Certificates
メスシリンダー、トールフォーム、PP
メスシリンダー、トールフォーム、PP
7 Options Available Below
Product Details & Documents
The 'nonwetting' polypropylene (PP) plastic surface of these single scale graduated cylinders eliminates formation of a concave meniscus and makes reading easier and more accurate. Chemically resistant, they are autoclavable at 121 °C (250 °F).
  • Single metric scale has raised numerals for easy reading
  • Flared top rims and large spouts facilitate pouring
  • Octagonal-shaped bases prevent tipping and rolling
Product Family Options
Product Information
  • 容量
    Division
    Availability
    Price
  • 10 ml
    0,2 ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $9.49
    $9.49
    1 * 1 items
       $9.49
  • 25 ml
    0,5 ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $9.74
    $9.74
    1 * 1 items
       $9.74
  • 50 ml
    1 ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $10.21
    $10.21
    1 * 1 items
       $10.21
  • 100 ml
    1 ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $12.89
    $12.89
    1 * 1 items
       $12.89
  • 250 ml
    2 ml
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $14.68
    $14.68
    1 * 1 items
       $14.68
  • 500 ml
    5 ml
    Log in for availability
    Log in for availability
    1 * 8 items
    1 * 8 items
  • 1000 ml
    10 ml
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items