Orders LinkContactUsLinkComponent
静電気防止用ブラシ、 StaticAway™ (スタティックアウェイ
静電気防止用ブラシ、 StaticAway™ (スタティックアウェイ
Supplier:   VWR Collection
静電気防止用ブラシ、 StaticAway™ (スタティックアウェイ
Supplier:  VWR Collection
Certificates
静電気防止用ブラシ、 StaticAway™ (スタティックアウェイ
静電気防止用ブラシ、 StaticAway™ (スタティックアウェイ
4 Options Available Below
Product Details & Documents
Brushes clean by dissipating static and whisking away harmful dust, lint, and dirt. Ideal for cleaning balance pans, optics, cuvettes, slides, film, microscopes, electronics, and computer monitors. The compact brush is suitable for electronic balances, providing accurate results by eliminating pan weight variances caused by static.
  • Natural bristles will not scratch critical surfaces
  • Brushes drain static and clean with a unique blend of natural hair, synthetic conductive fiber, and a conductive composite handle
  • Long handle brushes allow access to difficult places
Product Family Options
Product Information
  • 説明
    Bristle material
    Handle material
    総長
    Availability
    Price
  • Long handle brush
    Natural bristle
    Plastic
    25,4 mm
    1 * 1 items
  • Long handle brush
    Natural bristle
    Plastic
    50,8 mm
    1 * 1 items
  • Compact Brush
    Natural bristle
    Plastic
    63,5 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $117.30
    $117.30
    1 * 1 items
       $117.30
  • Long handle brush
    Natural bristle
    Plastic
    76,2 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - $89.55
    $89.55
    1 * 1 items
       $89.55