VWR® Basic Cleanroom Overalls, SMS
These cleanroom overalls are manufactured from a latex-free, multi-layer spun-bonded - meltblown - spun-bonded (SMS) fabric, that offers high tensile strength while remaining soft, comfortable and breathable.
- Tapered collar, inset sleeves, zip closure, elasticated cuffs
- Fluid and particulate barrier
- Light chemical splash resistance
These cleanroom overalls are manufactured from a latex-free, multi-layer spun-bonded - meltblown - spun-bonded (SMS) fabric, that offers high tensile strength while remaining soft, comfortable and breathable.
- Tapered collar, inset sleeves, zip closure, elasticated cuffs
- Fluid and particulate barrier
- Light chemical splash resistance
- Catalog No:
- 113-1097
- 113-1098
- 113-1099
- 113-1101
- 113-1102
- 113-1103
- 113-1090
- 113-1091
- 113-1092
- 113-1093
- 113-1094
- 113-1095
- 113-1096
- Garment application:
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
Product Family Options
- 説明Colour衣服のサイズ
Specifications
Cat. No.113-1097Garment applicationCleanroom|LaboratorySpecifications
Cat. No.113-1098Garment applicationCleanroom|LaboratorySpecifications
Cat. No.113-1099Garment applicationCleanroom|LaboratorySpecifications
Cat. No.113-1101Garment applicationCleanroom|LaboratorySpecifications
Cat. No.113-1102Garment applicationCleanroom|LaboratorySpecifications
Cat. No.113-1103Garment applicationCleanroom|LaboratorySpecifications
Cat. No.113-1090Garment applicationCleanroom|LaboratorySpecifications
Cat. No.113-1091Garment applicationCleanroom|LaboratorySpecifications
Cat. No.113-1092Garment applicationCleanroom|LaboratorySpecifications
Cat. No.113-1093Garment applicationCleanroom|LaboratorySpecifications
Cat. No.113-1094Garment applicationCleanroom|LaboratorySpecifications
Cat. No.113-1095Garment applicationCleanroom|LaboratorySpecifications
Cat. No.113-1096Garment applicationCleanroom|Laboratory