Orders LinkContactUsLinkComponent
VWR®, ブロックウェイト、クラスM1
Supplier:   VWR Collection
VWR®, ブロックウェイト、クラスM1
Supplier:  VWR Collection
Certificates
VWR®, ブロックウェイト、クラスM1
VWR®, ブロックウェイト、クラスM1
8 Options Available Below
Product Details & Documents
Rectangular weights made from cast iron (density 7,2 g/cm³).
  • Indelible unique identification number guarantees traceability
Colour: Grey.
Conforms to OILM, COFRAC accredited
 : Conforms to OIML, COFRAC accredited.
 : Block weights are supplied with cardboard packaging to protect the weights during transportation.
Product Family Options
Product Information
  • Weight
    Tolerance
    Type
    Availability
    Price
  • 5 kg
    250 ± mg
    Without certificate
    1 * 1 items
  • 10 kg
    500 ± mg
    Without certificate
    1 * 1 items
  • 20 kg
    1000 ± mg
    Without certificate
    1 * 1 items
  • 50 kg
    2500 ± mg
    Without certificate
    1 * 1 items
  • 5 kg
    250 ± mg
    With certificate
    1 * 1 items
  • 10 kg
    500 ± mg
    With certificate
    1 * 1 items
  • 20 kg
    1000 ± mg
    With certificate
    1 * 1 items
  • 50 kg
    2500 ± mg
    With certificate